SY3H3D67AF4E Synthonix Corporation

3-Chloro-4-morpholinobenzaldehyde

  • Cas Number : 886500-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54518.html

Safety Information

MSDS https://synthonix.com/54518.html
Availability details for
Quantity
  • Availability
  • Add to Cart