SY3H3D67AF50 Synthonix Corporation

(3R,4S)-4-(3-Chlorophenyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD11520170
  • Cas Number : 1047651-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54520.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54520.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart