SY3H3D67AF51 Synthonix Corporation

trans-4-(2-Chlorophenyl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD06659260
  • Cas Number : 1820581-20-1
  • Molecular Weight : 262.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/54521.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/54521.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart