SY3H3D67AF58 Synthonix Corporation

N-(6-Chloro-3-formylpyridin-2-yl)pivalamide

  • Cas Number : 127446-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54610.html

Safety Information

MSDS https://synthonix.com/54610.html
Availability details for
Quantity
  • Availability
  • Add to Cart