SY3H3D67AF5C Synthonix Corporation

2-(4-Chlorophenyl)-3-methylbutanoic acid

  • Cas Number : 2012-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54676.html

Safety Information

MSDS https://synthonix.com/54676.html
Availability details for
Quantity
  • Availability
  • Add to Cart