SY3H3D67AF65 Synthonix Corporation

1-Cyclopropyl-1-phenylethanol

  • MDL number MFCD00019243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54759.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54759.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart