SY3H3D67AF69 Synthonix Corporation

1-Cyclopropyl-1-(4-methoxyphenyl)methylamine

  • MDL number MFCD00044830
  • Cas Number : 54398-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54859.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54859.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart