SY3H3D67AF7D Synthonix Corporation

4-Chloro-8-methoxyquinoline-3-carbonitrile

  • Cas Number : 214476-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55188.html

Safety Information

MSDS https://synthonix.com/55188.html
Availability details for
Quantity
  • Availability
  • Add to Cart