SY3H3D67AF87 Synthonix Corporation

4-Chloro-1-methyl-2-oxo-1,2-dihydroquinoline-3-carbaldehyde

  • MDL number MFCD00204024
  • Cas Number : 96600-76-9
  • Molecular Weight : 221.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-1-methyl-2-oxo-1,2-dihydro-3-quinolinecarbaldehyde
Inchi Code 1S/C11H8ClNO2/c1-13-9-5-3-2-4-7(9)10(12)8(6-14)11(13)15/h2-6H,1H3
Inchi Key HXFNBXZHPBNABA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Yellow Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/55277.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/55277.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart