SY3H3D67AF89 Synthonix Corporation

(4-Chlorophenyl)(1H-pyrrol-3-yl)methanone

  • Cas Number : 62128-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55289.html

Safety Information

MSDS https://synthonix.com/55289.html
Availability details for
Quantity
  • Availability
  • Add to Cart