SY3H3D67AF8C Synthonix Corporation

2-Chloro-4-methyl-6-phenylpyrimidine

  • Cas Number : 32785-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55312.html

Safety Information

MSDS https://synthonix.com/55312.html
Availability details for
Quantity
  • Availability
  • Add to Cart