SY3H3D67AF91 Synthonix Corporation

5-(4-Chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08144097
  • Cas Number : 957513-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55320.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55320.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart