SY3H3D67AF93 Synthonix Corporation

4-Chloro-6-methoxy-2-phenylpyrimidine

  • Cas Number : 4319-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55331.html

Safety Information

MSDS https://synthonix.com/55331.html
Availability details for
Quantity
  • Availability
  • Add to Cart