SY3H3D67AF95 Synthonix Corporation

Cyclobutyl(phenyl)methanamine hydrochloride

  • Cas Number : 58271-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55481.html

Safety Information

MSDS https://synthonix.com/55481.html
Availability details for
Quantity
  • Availability
  • Add to Cart