SY3H3D67AF9A Synthonix Corporation

(R)-Cyclopropyl(3-methoxyphenyl)methanamine hydrochloride

  • Cas Number : 1391455-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55500.html

Safety Information

MSDS https://synthonix.com/55500.html
Availability details for
Quantity
  • Availability
  • Add to Cart