SY3H3D67AFA4 Synthonix Corporation

(4-Chlorophenyl)(pyridin-2-yl)methanol

  • Cas Number : 27652-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55603.html

Safety Information

MSDS https://synthonix.com/55603.html
Availability details for
Quantity
  • Availability
  • Add to Cart