SY3H3D67AFA5 Synthonix Corporation

4-(Cyclopropylethynyl)benzoic acid

  • Cas Number : 908247-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55612.html

Safety Information

MSDS https://synthonix.com/55612.html
Availability details for
Quantity
  • Availability
  • Add to Cart