SY3H3D67AFA6 Synthonix Corporation

(4'-Chloro-[1,1'-biphenyl]-4-yl)boronic acid

  • MDL number MFCD08544389
  • Cas Number : 364044-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55614.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55614.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart