SY3H3D67AFB4 Synthonix Corporation

N-Cbz-D-Asparagine

  • MDL number MFCD00065696
  • Cas Number : 4474-86-6
  • Molecular Weight : 266.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55881.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55881.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart