SY3H3D67AFBD Synthonix Corporation

Cyclopentyl(phenyl)methanone

  • MDL number MFCD00001378
  • Cas Number : 5422-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55913.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55913.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart