SY3H3D67AFC0 Synthonix Corporation

1-(2-Chloro-4-morpholinothieno[3,2-d]pyrimidin-6-yl)-N-methylmethanamine

  • MDL number MFCD28966047
  • Cas Number : 955979-15-4
  • Molecular Weight : 298.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55970.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55970.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart