SY3H3D67AFC3 Synthonix Corporation

2-(5-Chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD16036133
  • Cas Number : 1190129-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56004.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56004.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart