SY3H3D67AFC7 Synthonix Corporation

1-Cyclopentyl-4-methylbenzene

  • Cas Number : 827-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56018.html

Safety Information

MSDS https://synthonix.com/56018.html
Availability details for
Quantity
  • Availability
  • Add to Cart