SY3H3D67AFC8 Synthonix Corporation

3-Cbz-Aminopyrrolidine

  • MDL number MFCD04115314
  • Cas Number : 115551-46-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Sticky Oil to Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56029.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56029.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart