SY3H3D67AFCA Synthonix Corporation

2-(3-(4-Chlorophenyl)pyrrolidin-3-yl)ethanol

  • MDL number MFCD17014882
  • Cas Number : 52423-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56067.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56067.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart