SY3H3D67AFDA Synthonix Corporation

5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • Cas Number : 863578-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56158.html

Safety Information

MSDS https://synthonix.com/56158.html
Availability details for
Quantity
  • Availability
  • Add to Cart