SY3H3D67AFE8 Synthonix Corporation

1-Chloro-5H-pyrido[4,3-b]indole-4-carboxamide

  • Cas Number : 938465-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56433.html

Safety Information

MSDS https://synthonix.com/56433.html
Availability details for
Quantity
  • Availability
  • Add to Cart