SY3H3D67AFE9 Synthonix Corporation

1-Chloro-9H-carbazole

  • MDL number MFCD18450171
  • Cas Number : 5599-70-2
  • Molecular Weight : 201.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to pale-red or yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56438.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56438.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart