SY3H3D67AFEC Synthonix Corporation

6-Chloro-5-phenylpicolinic acid

  • Cas Number : 1214352-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56446.html

Safety Information

MSDS https://synthonix.com/56446.html
Availability details for
Quantity
  • Availability
  • Add to Cart