SY3H3D67AFFC Synthonix Corporation

(S)-1-Cbz-3-Aminopyrrolidine hydrochloride

  • Cas Number : 550378-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56612.html

Safety Information

MSDS https://synthonix.com/56612.html
Availability details for
Quantity
  • Availability
  • Add to Cart