SY3H3D67B003 Synthonix Corporation

4-Chloro-4'-methyl-1,1'-biphenyl

  • Cas Number : 19482-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56695.html

Safety Information

MSDS https://synthonix.com/56695.html
Availability details for
Quantity
  • Availability
  • Add to Cart