SY3H3D67B004 Synthonix Corporation

(4-(Chloromethyl)phenyl)(phenyl)sulfane

  • Cas Number : 1208-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56696.html

Safety Information

MSDS https://synthonix.com/56696.html
Availability details for
Quantity
  • Availability
  • Add to Cart