SY3H3D67B00F Synthonix Corporation

(3-(4-Chlorophenoxy)phenyl)methanamine hydrochloride

  • Cas Number : 1171023-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56768.html

Safety Information

MSDS https://synthonix.com/56768.html
Availability details for
Quantity
  • Availability
  • Add to Cart