SY3H3D67B013 Synthonix Corporation

4-Cyano-4-((phenylcarbonothioyl)thio)pentanoic acid

  • MDL number MFCD10698690
  • Cas Number : 201611-92-9
  • Molecular Weight : 279.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56824.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/56824.html
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart