SY3H3D67B015 Synthonix Corporation

(2S,4R)-4-(2-Cyanobenzyl)pyrrolidine-2-carboxylic acid

  • Cas Number : 1049981-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56849.html

Safety Information

MSDS https://synthonix.com/56849.html
Availability details for
Quantity
  • Availability
  • Add to Cart