SY3H3D67B019 Synthonix Corporation

3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number No MDL found
  • Cas Number : 945391-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56911.html

Safety Information

MSDS https://synthonix.com/56911.html
Availability details for
Quantity
  • Availability
  • Add to Cart