SY3H3D67B01A Synthonix Corporation

N-Cbz-D-glutamic Acid

  • MDL number MFCD00063193
  • Cas Number : 63648-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56933.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56933.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart