SY3H3D67B01D Synthonix Corporation

2-cyclopentyl-2-hydroxy-2-phenylacetic acid

  • MDL number MFCD00019296
  • Cas Number : 427-49-6
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56983.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56983.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart