SY3H3D67B01F Synthonix Corporation

4-Cbz-Aminopiperidine

  • Cas Number : 182223-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57069.html

Safety Information

MSDS https://synthonix.com/57069.html
Availability details for
Quantity
  • Availability
  • Add to Cart