SY3H3D67B020 Synthonix Corporation

(R)-1-Cbz-2-methylpiperazine

  • MDL number MFCD04115051
  • Cas Number : 923565-99-5
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Colorless to Yellow Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57070.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57070.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart