SY3H3D67B022 Synthonix Corporation

2-(3-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 517920-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57119.html

Safety Information

MSDS https://synthonix.com/57119.html
Availability details for
Quantity
  • Availability
  • Add to Cart