SY3H3D67B03A Synthonix Corporation

4-Chloro-5-(p-tolyl)thieno[2,3-d]pyrimidine

  • MDL number MFCD01238585
  • Cas Number : 374104-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57345.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57345.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart