SY3H3D67B03B Synthonix Corporation

2-Chloro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD16875829
  • Cas Number : 1227268-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57348.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57348.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart