SY3H3D67B03E Synthonix Corporation

2-Chloro-1-((3-fluorobenzyl)oxy)-4-nitrobenzene

  • MDL number MFCD06809827
  • Cas Number : 443882-99-3
  • Molecular Weight : 281.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57368.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57368.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart