SY3H3D67B045 Synthonix Corporation

4'-Chloro-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD01631911
  • Cas Number : 80565-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57388.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57388.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart