SY3H3D67B04E Synthonix Corporation

3-((2-Chloro-4-nitrophenoxy)methyl)benzoic acid

  • Cas Number : 30880-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57489.html

Safety Information

MSDS https://synthonix.com/57489.html
Availability details for
Quantity
  • Availability
  • Add to Cart