SY3H3D67B05F Synthonix Corporation

4-Chloro-1-(2-methylpropyl)-1H-imidazo[4,5-c]quinoline

  • MDL number MFCD06658174
  • Cas Number : 99010-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57640.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57640.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart