SY3H3D67B060 Synthonix Corporation

N-(2-Chloropyrimidin-4-yl)-N,2,3-trimethyl-2H-indazol-6-amine

  • Cas Number : 444731-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57643.html

Safety Information

MSDS https://synthonix.com/57643.html
Availability details for
Quantity
  • Availability
  • Add to Cart