SY3H3D67B061 Synthonix Corporation

3-(2-Chloro-2-(4-chlorophenyl)vinyl)-2,2-dimethylcyclopropanecarboxylic acid

  • MDL number MFCD28048697
  • Cas Number : 88419-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57656.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57656.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart