SY3H3D67B06B Synthonix Corporation

(S)-3-(Cbz-amino)-2-oxoazepane

  • MDL number MFCD08060107
  • Cas Number : 103478-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57772.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57772.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart